(2S)-2-Decanamidopropanoic acid 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: N-Decanoyl-L-alanine
Formula:C₁₃H₂₅NO₃
MW:243.34 g/mol
CAS Number:110538-83-5
:(2S)-2-Decanamidopropanoic acid
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 243.34 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:95%
- Appearance:white powder
- (2S)-2-Decanamidopropanoic acid 95%:
Specification Test Results
Customers Who Bought This Also Bought