Hydroxy-PEG2-methylamine 97%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: 2-(2-(2-(methylamino)ethoxy)ethoxy)ethan-1-ol
Formula:C₇H₁₇NO₃
MW:163.22 g/mol
MDL Number:MFCD30182052
CAS Number:282551-10-4
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97%
:97
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 163.22 g/mol
:Hydroxy-PEG2-methylamine
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:97%
- Appearance:Clear Oil
- Hydroxy-PEG2-methylamine 97%:
Specification Test Results
Customers Who Bought This Also Bought