2-[2-(trifluoroacetamido)acetamido]acetic acid 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Formula:C₆H₇F₃N₂O₄
MW:228.13 g/mol
CAS Number:400-58-8
:2-[2-(trifluoroacetamido)acetamido]acetic acid
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 228.13 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:95%
- 2-[2-(trifluoroacetamido)acetamido]acetic acid 95%:
Specification Test Results
Customers Who Bought This Also Bought