2,4,7,8,9-Penta-O-acetyl-N-acetylneuraminic acid methyl ester 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: (1S,2R)-1-((2R,3R,4S)-3-Acetamido-4,6-diacetoxy-6-(methoxycarbonyl)tetrahydro-2H-pyran-2-yl)propane-1,2,3-triyl triacetate,N-Acetylneuraminic acid methyl ester 2,4,7,8,9-Pentaacetate,Methyl 5-Acetamido-2,4,7,8,9-penta-O-acetyl-3
CAS Number:73208-82-9
:MFCD11112172
Formula:C₂₂H₃₁NO₁₄
MW:533.48 g/mol
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 533.48 g/mol
:2,4,7,8,9-Penta-O-acetyl-N-acetylneuraminic acid methyl ester
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:95%
- Appearance:white powder
- 2,4,7,8,9-Penta-O-acetyl-N-acetylneuraminic acid methyl ester 95%:
Specification Test Results
Customers Who Bought This Also Bought