Benzyl-2-acetamido-2-deoxy-α-D-galactopyranoside 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Formula:C₁₅H₂₁NO₆
MW:311.33 g/mol
MDL Number:MFCD00057920
CAS Number:3554-93-6
:Benzyl-2-acetamido-2-deoxy-α-D-galactopyranoside
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 311.33 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:95%
- Appearance:white powder
- Benzyl-2-acetamido-2-deoxy-α-D-galactopyranoside 95%:
Specification Test Results
Customers Who Bought This Also Bought