Allyl 2,3,4,6-tetra-O-acetyl-β-D-glucopyranoside 97%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: Allyl-tetra-O-acetyl-β-D-glucopyranoside
Formula:C₁₇H₂₄O₁₀
MW:388.37 g/mol
MDL Number:MFCD03427087
CAS Number:10343-15-4
:Allyl 2,3,4,6-tetra-O-acetyl-β-D-glucopyranoside
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97%
:97
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 388.37 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:97%
- Appearance:White Solid
- Allyl 2,3,4,6-tetra-O-acetyl-β-D-glucopyranoside 97%:
Specification Test Results
Customers Who Bought This Also Bought