N-acetyl-L-fucosamine ≥95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: 2-Acetamido-2
Formula:C₈H₁₅NO₅
MW:205.21 g/mol
MDL Number:MFCD22124196
CAS Number:49694-69-1
:N-acetyl-L-fucosamine
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 205.21 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:> 95%
- Appearance:white powder
- N-acetyl-L-fucosamine ≥95%:
Specification Test Results
Customers Who Bought This Also Bought