2,3,4,6-Tetra-O-acetyl-D-galactopyranose 97%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Formula:C₁₄H₂₀O₁₀
MW:348.3 g/mol
MDL Number:MFCD09817577
CAS Number:47339-09-3
:2,3,4,6-Tetra-O-acetyl-D-galactopyranose
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97%
:97
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 348.3 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:97%
- Appearance:White powder
- 2,3,4,6-Tetra-O-acetyl-D-galactopyranose 97%:
Specification Test Results
Customers Who Bought This Also Bought