β-GlcNAc-PEG3-Amine 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: 2-{2-[2-aminoethoxy]ethoxy}ethyl 2-acetamido-2-deoxy-β-D-galactopyranoside
Formula:C₁₄H₂₈N₂O₈
MW:352.38 g/mol
CAS Number:212896-46-3
:β-GlcNAc-PEG3-Amine
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 352.38 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:95%
- Appearance:colorless sticky syrup
- β-GlcNAc-PEG3-Amine 95%:
Specification Test Results
Customers Who Bought This Also Bought