2-propyn-1-yl 2-(acetylamino) α-D-Galactoside ≥95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: prop-2-yn-1-yl 2-acetamido-2-deoxy-alpha-D-galactopyranoside
Formula:C₁₁H₁₇NO₆
MW:259.26 g/mol
CAS Number:1152423-03-4
:2-propyn-1-yl 2-(acetylamino) α-D-Galactoside
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 259.26 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:> 95%
- Appearance:White Powder
- 2-propyn-1-yl 2-(acetylamino) α-D-Galactoside ≥95%:
Specification Test Results
Customers Who Bought This Also Bought