2,4,7,8,9-Penta-O-acetyl-N-azidoacetyl-neuraminic acid methyl ester ≥95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: Ac5NeuNAz-O-Me
Formula:C₂₂H₃₀N₄O₁₄
MW:574.5 g/mol
CAS Number:1357804-21-7
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 574.5 g/mol
:2,4,7,8,9-Penta-O-acetyl-N-azidoacetyl-neuraminic acid methyl ester
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:> 95%
- Appearance:white powder
- 2,4,7,8,9-Penta-O-acetyl-N-azidoacetyl-neuraminic acid methyl ester ≥95%:
Specification Test Results
Customers Who Bought This Also Bought