2,3,6,2',3',4',6'-Hepta-O-acetyl-b-maltosyl azide 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: 2,3,6-Tri-O-acetyl-4-O-(2,3,4,6-tetra-O-acetyl-a-D-glucopyranosyl)-b-D-glucopyranosyl azide
Formula:C₂₆H₃₅N₃O₁₇
MW:661.57 g/mol
MDL Number:MFCD00216969
CAS Number:33012-49-6
:2,3,6,2',3',4',6'-Hepta-O-acetyl-b-maltosyl azide
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 661.57 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:95%
- Appearance:Off-white powder
- 2,3,6,2',3',4',6'-Hepta-O-acetyl-b-maltosyl azide 95%:
Specification Test Results
Customers Who Bought This Also Bought