1,3,4-Tri-O-acetyl-2-azido-2-deoxy-α-L-fucopyranose ≥95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: 2-Azido-2,6-dideoxy-α-L-galactopyranose 1,3
Formula:C₁₂H₁₇N₃O₇
MW:315.28 g/mol
CAS Number:115435-17-1
:1,3,4-Tri-O-acetyl-2-azido-2-deoxy-α-L-fucopyranose
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 315.28 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:> 95%
- Appearance:white powder
- 1,3,4-Tri-O-acetyl-2-azido-2-deoxy-α-L-fucopyranose ≥95%:
Specification Test Results
Customers Who Bought This Also Bought