6-Azido-6-deoxy-1,2:3,4-di-O-isopropylidene-a-D-galactopyranose 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Formula:C₁₂H₁₉N₃O₅
MW:285.3 g/mol
MDL Number:MFCD02683313
CAS Number:4711-00-6
:6-Azido-6-deoxy-1,2:3,4-di-O-isopropylidene-a-D-galactopyranose
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 285.3 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:95%
- Appearance:Colorless Syrup
- 6-Azido-6-deoxy-1,2:3,4-di-O-isopropylidene-a-D-galactopyranose 95%:
Specification Test Results
Customers Who Bought This Also Bought