2,3,4,6-Tetra-O-acetyl-β-D-galactopyranosyl PEG3-azide ≥95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: β-Gal-PEG3-azide tetraacetate
Formula:C₂₀H₃₁N₃O₁₂
MW:505.5 g/mol
MDL Number:MFCD15145111
CAS Number:126765-25-1
:2,3,4,6-Tetra-O-acetyl-β-D-galactopyranosyl PEG3-azide
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 505.5 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:> 95%
- Appearance:Colorless sticky oil
- 2,3,4,6-Tetra-O-acetyl-β-D-galactopyranosyl PEG3-azide ≥95%:
Specification Test Results
Customers Who Bought This Also Bought