5-Azidopentyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-galactopyranoside 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Formula:C₁₉H₃₀N₄O₉
MW:458.47 g/mol
CAS Number:1479084-10-0
:5-Azidopentyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-galactopyranoside
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 458.47 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:95%
- Appearance:colorless sticky oil
- 5-Azidopentyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-galactopyranoside 95%:
Specification Test Results
Customers Who Bought This Also Bought