β-D-GlcNAc-PEG3-Azide, 3,4,6-triacetate 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: (2-(2-(2-Azidoethoxy)ethoxy)ethyl)-2-acetamido-2-deoxy-3,4
Formula:C₂₀H₃₂N₄O₁₁
MW:504.49 g/mol
CAS Number:86520-53-8
:β-D-GlcNAc-PEG3-Azide, 3,4,6-triacetate
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 504.49 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:95%
- β-D-GlcNAc-PEG3-Azide, 3,4,6-triacetate 95%:
Specification Test Results
Customers Who Bought This Also Bought