Hepta-O-acetyl-β-lactosyl azide 98%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: 2,3,6-Tri-O-acetyl-4-O-(2,3,4
Formula:C₂₆H₃₅N₃O₁₇
MW:661.57 g/mol
MDL Number:MFCD05664728
CAS Number:30854-62-7
:Hepta-O-acetyl-β-lactosyl azide
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98%
:98
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 661.57 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:98%
- Appearance:White powder
- Hepta-O-acetyl-β-lactosyl azide 98%:
Specification Test Results
Customers Who Bought This Also Bought