alpha-GalNAc-PEG3-Azide ≥95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: 2-{2-[2-azidoethoxy]ethoxy}ethyl 2-acetamido-2-deoxy-α-D-galactopyranoside
Formula:C₁₄H₂₆N₄O₈
MW:378.38 g/mol
CAS Number:882873-70-3
:alpha-GalNAc-PEG3-Azide
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 378.38 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:> 95%
- Appearance:White powder
- alpha-GalNAc-PEG3-Azide ≥95%:
Specification Test Results
Customers Who Bought This Also Bought