Methyl-2,3,4-tri-O-acetyl-L-fucopyranoside 97%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: Methyl 6-deoxy-L-galactopyranoside triacetate
Formula:C₁₃H₂₀O₈
MW:304.29 g/mol
MDL Number:MFCD27665349
CAS Number:67576-78-7
:Methyl-2,3,4-tri-O-acetyl-L-fucopyranoside
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97%
:97
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 304.29 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:97%
- Appearance:White powder
- Methyl-2,3,4-tri-O-acetyl-L-fucopyranoside 97%:
Specification Test Results
Customers Who Bought This Also Bought