TBS-DHG Catalyst ≥95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: (2R,3S,4R)-2-(((tert-Butyldimethylsilyl)oxy)methyl)tetrahydro-2H-pyran-3,4-diol, 6-Tertbutyldimethylsilyl-1
Formula:C₁₂H₂₆O₄Si
MW:262.42 g/mol
CAS Number:212120-61-1
:TBS-DHG Catalyst
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 262.42 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:> 95%
- Appearance:Powder or Crystals
- TBS-DHG Catalyst ≥95%:
Specification Test Results
Customers Who Bought This Also Bought