Phenyl 2,3,4,6-tetra-O-acetyl-α-D-thiomannopyranoside 97%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Formula:C₂₀H₂₄O₉S
MW:440.46 g/mol
MDL Number:MFCD01862644
CAS Number:13992-16-0
:Phenyl 2,3,4,6-tetra-O-acetyl-α-D-thiomannopyranoside
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97%
:97
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 440.46 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:97%
- Appearance:white powder
- Phenyl 2,3,4,6-tetra-O-acetyl-α-D-thiomannopyranoside 97%:
Specification Test Results
Customers Who Bought This Also Bought