Methyl 2,3,4-tri-O-acetyl-6-O-trityl-α-D-galactopyranoside 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Formula:C₃₂H₃₄O₉
MW:562.61 g/mol
MDL Number:MFCD09952885
CAS Number:38982-56-8
:Methyl 2,3,4-tri-O-acetyl-6-O-trityl-α-D-galactopyranoside
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 562.61 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:95%
- Appearance:White solid
- Methyl 2,3,4-tri-O-acetyl-6-O-trityl-α-D-galactopyranoside 95%:
Specification Test Results
Customers Who Bought This Also Bought