6-O-tert-Butyldimethylsilyl-D-galactal 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Formula:C₁₂H₂₄O₄Si
MW:260.4 g/mol
MDL Number:MFCD09756159
CAS Number:124751-19-5
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 260.4 g/mol
:6-O-tert-Butyldimethylsilyl-D-galactal
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:95%
- Appearance:Colorless Syrup
- 6-O-tert-Butyldimethylsilyl-D-galactal 95%:
Specification Test Results
Customers Who Bought This Also Bought