6-O-tert-Butyldiphenylsilyl-D-galactal ≥95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: (2R,3R,4R)-2-(((tert-Butyldiphenylsilyl)oxy)methyl)-3,4-dihydro-2H-pyran-3
Formula:C₂₂H₂₈O₄Si
MW:384.55 g/mol
CAS Number:137893-35-7
:6-O-tert-Butyldiphenylsilyl-D-galactal
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 384.55 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:> 95%
- Appearance:Sticky Syrup
- 6-O-tert-Butyldiphenylsilyl-D-galactal ≥95%:
Specification Test Results
Customers Who Bought This Also Bought