Fmoc-L-Ser(β-D-Gal(Ac)4)-OH 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: N-α-(9-fluorenylmethyloxycarbonyl)-O-β-(2,3,4
Formula:C₃₂H₃₅NO₁₄
MW:657.62 g/mol
MDL Number:MFCD05663657
CAS Number:96383-44-7
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 657.62 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:95%
- Appearance:White powder
- Fmoc-L-Ser(β-D-Gal(Ac)4)-OH 95%:
Specification Test Results
Customers Who Bought This Also Bought