Ethyl 2-deoxy-2-(1,3-dihydro-1,3-dioxo-2H-isoindol-2-yl)-1-thio-beta-D-glucopyranoside 98% (by HPLC)
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Formula:C₁₆H₁₉NO₆S
MW:353.39 g/mol
MDL Number:MFCD28167068
CAS Number:130539-43-4
:Ethyl 2-deoxy-2-(1,3-dihydro-1,3-dioxo-2H-isoindol-2-yl)-1-thio-beta-D-glucopyranoside
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98%
:98
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 353.39 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:98% (HPLC)
- Appearance:Off-white powder
- Ethyl 2-deoxy-2-(1,3-dihydro-1,3-dioxo-2H-isoindol-2-yl)-1-thio-beta-D-glucopyranoside 98%(by HPLC):
Specification Test Results
Customers Who Bought This Also Bought