Ethyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-thiogalactopyranoside 98% (by HPLC)
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: β-D-Galactopyranoside, ethyl 2-(acetylamino)-2-deoxy-1-thio-, 3,4,6-triacetate,(2R,3R,4R,5R,6S)-5-Acetamido-2-(acetoxymethyl)-6-(ethylthio)tetrahydro-2H-pyran-3
Formula:C₁₆H₂₅NO₈S
MW:391.436 g/mol
MDL Number:MFCD31814466
CAS Number:144218-98-4
:Ethyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-thiogalactopyranoside
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98%
:98
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 391.436 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:98% (HPLC)
- Appearance:Off-white powder
- Ethyl 2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-thiogalactopyranoside 98%(by HPLC):
Specification Test Results
Customers Who Bought This Also Bought