tert-Butyl (2-(2-(2-(2-bromoacetamido)ethoxy)ethoxy)ethyl)carbamate
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: Bromoacetamido-PEG2-Boc-amine
Formula:C₁₃H₂₅BrN₂O₅
MW:369.252 g/mol
MDL Number:MFCD28142467
CAS Number:182244-33-3
:tert-Butyl (2-(2-(2-(2-bromoacetamido)ethoxy)ethoxy)ethyl)carbamate
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 369.252 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- tert-Butyl (2-(2-(2-(2-bromoacetamido)ethoxy)ethoxy)ethyl)carbamate:
Specification Test Results
Customers Who Bought This Also Bought