1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-D-galactopyranose
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: 2-Azido-2-deoxy-D-galactose peracetate,1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-D-galactopyranose,2-Azido-2-deoxy-D-galactopyranose 1,3,4
Formula:C₁₄H₁₉N₃O₉
MW:373.315 g/mol
MDL Number:MFCD02683262
CAS Number:84278-00-2
:1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-D-galactopyranose
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 373.315 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- 1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-D-galactopyranose:
Specification Test Results
Customers Who Bought This Also Bought