2,3,4,6-Tetra-O-acetyl-α-D-mannopyranosyl azide 97%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Formula:C₁₄H₁₉N₃O₉
MW:373.315 g/mol
MDL Number:MFCD05664724
CAS Number:53784-29-5
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97%
:97
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 373.315 g/mol
:2,3,4,6-Tetra-O-acetyl-α-D-mannopyranosyl azide
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:97%
- Appearance:White powder
- 2,3,4,6-Tetra-O-acetyl-α-D-mannopyranosyl azide 97%:
Specification Test Results
Customers Who Bought This Also Bought