2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-galactopyranose ≥95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: (2S,3R,4R,5R,6R)-3-Acetamido-6-(acetoxymethyl)tetrahydro-2H-pyran-2,4,5-triyl triacetate,β-GalNAc tetraacetate,b-D-Galactosamine pentaacetate,1,3,4,6-Tetra-O-acetyl-β-GalNAc,1,3,4,6-Tetra-O-acetyl-2-(acetylamino)-2-deoxy-β-D-galactopyranose
Formula:C₁₆H₂₃NO₁₀
MW:389.355 g/mol
MDL Number:MFCD00214297
CAS Number:3006-60-8
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 389.355 g/mol
:2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-galactopyranose
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:> 95%
- Appearance:white Solid
- 2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-β-D-galactopyranose ≥95%:
Specification Test Results
Customers Who Bought This Also Bought