Phenylethyl-β-D-thiogalactopyranoside ≥98%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(phenethylthio)tetrahydro-2H-pyran-3,4
Formula:C₁₄H₂₀O₅S
MW:300.3706 g/mol
MDL Number:MFCD00467904
CAS Number:63407-54-5
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 300.3706 g/mol
:Phenylethyl-β-D-thiogalactopyranoside
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:> 98%
- Appearance:White Powder
- Phenylethyl-β-D-thiogalactopyranoside ≥98%:
Specification Test Results
Customers Who Bought This Also Bought