Gastrodin ≥97%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: 4-(Hydroxymethyl)phenyl beta-D-Glucopyranoside,(2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-(hydroxymethyl)phenoxy)tetrahydro-2H-pyran-3,4
Formula:C₁₃H₁₈O₇
MW:286.28 g/mol
MDL Number:MFCD00272169
CAS Number:62499-27-8
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 286.28 g/mol
:Gastrodin
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:> 97%
- Appearance:White Powder
- Gastrodin ≥97%:
Specification Test Results
Customers Who Bought This Also Bought