2-Azidoethyl 2-acetamido-2-deoxy-β-D-galactopyranoside 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: β-D-GalNAc-Et-N3
Formula:C₁₀H₁₈N₄O₆
MW:290.28 g/mol
MDL Number:MFCD19981043
CAS Number:142072-15-9
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 290.28 g/mol
:2-Azidoethyl 2-acetamido-2-deoxy-β-D-galactopyranoside
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:95%
- Appearance:White powder
- 2-Azidoethyl 2-acetamido-2-deoxy-β-D-galactopyranoside 95%:
Specification Test Results
Customers Who Bought This Also Bought