3,4,6-Tri-O-benzyl-D-galactal 98%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: Tri-O-Benzyl-D-galactal
Formula:C₂₇H₂₈O₄
MW:416.517 g/mol
MDL Number:MFCD09039276
CAS Number:80040-79-5
:3,4,6-Tri-O-benzyl-D-galactal
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98%
:98
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 416.517 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:98%
- Appearance:White solid
- 3,4,6-Tri-O-benzyl-D-galactal 98%:
Specification Test Results
Customers Who Bought This Also Bought