Fmoc-L-Ser(α-D-GalNAc(Ac)3)-OH ≥95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: Fmoc-L-Ser (α-D-GalNAc(Ac)3)-OH,N-α-Fmoc-O-β-(2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-galactopyranosyl)-L-serine,Fmoc-Ser(GalNAc(Ac)₃-α-D)-OH
Formula:C₃₂H₃₆N₂O₁₃
MW:656.641 g/mol
MDL Number:MFCD00171375
CAS Number:120173-57-1
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 656.641 g/mol
:Fmoc-L-Ser(α-D-GalNAc(Ac)3)-OH
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:> 95%
- Appearance:White Powder
- Fmoc-L-Ser(α-D-GalNAc(Ac)3)-OH ≥95%:
Specification Test Results
Customers Who Bought This Also Bought