Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside ≥97%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: 1-Phenyl-1-thio-β-D-glucopyranoside tetraacetate,(2R,3R,4S,5R,6S)-2-(Acetoxymethyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triyl triacetate,Phenyl 2,3,4
Formula:C₂₀H₂₄O₉S
MW:440.46 g/mol
MDL Number:MFCD00135159
CAS Number:23661-28-1
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 440.46 g/mol
:Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:> 97%
- Appearance:White Powder
- Phenyl-2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside ≥97%:
Specification Test Results
Customers Who Bought This Also Bought