4,7,8,9-Tetra-O-acetyl-N-acetylneuraminic acid methyl ester ≥95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: Methyl 5-Acetamido-4,7,8,9-tetra-O-acetyl-3,5-dideoxy-D-glycero-D-galacto-2-nonulopyranosonate,Methyl 4,7,8,9-tetra-O-acetyl-N-acetylneuraminate,N-Acetylneuraminic acid methyl ester 4,7,8
Formula:C₂₀H₂₉NO₁₃
MW:491.45 g/mol
MDL Number:MFCD08276266
CAS Number:84380-10-9
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 491.45 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:> 95%
- Appearance:White Powder
- 4,7,8,9-Tetra-O-acetyl-N-acetylneuraminic acid methyl ester ≥95%:
Specification Test Results
Customers Who Bought This Also Bought