2-Azidoethyl-β-D-glucopyranoside 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: β-D-Glc-Et-N3
Formula:C₈H₁₅N₃O₆
MW:249.22 g/mol
CAS Number:165331-08-8
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 249.22 g/mol
:2-Azidoethyl-β-D-glucopyranoside
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:95%
- Appearance:Colorless Syrup
- 2-Azidoethyl-β-D-glucopyranoside 95%:
Specification Test Results
Customers Who Bought This Also Bought