2,3,4,6-Tetra-O-acetyl-ɑ-D-galactopyranosyl-2,2,2-trichloroacetimidate ≥95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: (2R,3S,4S,5R,6R)-2-(Acetoxymethyl)-6-(2,2,2-trichloro-1-iminoethoxy)tetrahydro-2H-pyran-3,4
Formula:C₁₆H₂₀Cl₃NO₁₀
MW:492.68 g/mol
MDL Number:MFCD07369652
CAS Number:86520-63-0
:Storage temperature: Refrigerator
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 492.68 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Purity Limit:> 95%
- Appearance:white powder
- 2,3,4,6-Tetra-O-acetyl-ɑ-D-galactopyranosyl-2,2,2-trichloroacetimidate ≥95%:
Specification Test Results
Customers Who Bought This Also Bought