α-GalNAc-PEG3-alkyne 95%
Supplier: AFG BIOSCIENCE LLC
Certificates
About this item
Synonyms: 2-{2-[2-(Prop-2-ynyloxy)ethoxy]ethoxy}ethyl 2-acetamido-2-deoxy-α-D-galactopyranoside
Formula:C₁₇H₂₉NO₉
MW:391.41 g/mol
:α-GalNAc-PEG3-alkyne
:Storage temperature: Freezer
:Length:
:Width:
:Height:
:Ø
:Thickness:
:95%
:95
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 391.41 g/mol
Storage Temperature:Freezer
Specification Test Results
- Purity Limit:95%
- Appearance:White powder
- α-GalNAc-PEG3-alkyne 95%:
Specification Test Results
Customers Who Bought This Also Bought