Specifications
- Description:Laser histology microscope slide printer; 100-slide cartridge capacity, up to 35-slide output tray capacity
- Cat. no.:76632-834
- Dimensions:46L×20W×34H cm (18.1×7.9×13.4")
- Weight:19 kg (42 lbs.)
Specifications
About this item
Azer Presto™ Microscope Slide Printer allows for quick, permanent and efficient printing on microscope slides. UV laser technology eliminates the need of expensive printer consumables and maintenance.
- Integrate to LIS/HIS system
- High resolution 500×500 dpi
- High speed; as fast as three seconds for on-demand printing
- Input cartridge holds up to 100 slides and output holds up to 35 slides
- Continuous and on-demanding printing model
- Barcode, QR code, logo, alphanumeric, graphic printing
- ≤65dB work noise
Azer Scientific's Presto™ microscope slide printer is an ultraviolet laser model that can integrate into any laboratory or hospital information system. Our UV laser technology creates a permanent labeling resistant to degradation. Implementing the new Azer Scientific Presto™ slide printer aids in a time management workflow that you will not want to resist.