MabSelect™ VL Protein L Resin, Cytiva
Catalog #: CA76633-028
Supplier: Cytiva
CAS Number:
Specifications
- Particle size:~60 µm
- pH range:2 - 13
- Description:VL protein L resin
- Storage conditions:2...8 °C, 20% ethanol
- Matrix:Highly cross-linked agarose, spherical
- Ligand:Alkaline stabilized protein L-derived (E. coli)
- Ligand coupling method:Epoxy
- Cat. no.:CA76633-028
- Volume:25 ml
Specifications
About this item
MabSelect™ VL resin with a redesigned protein L ligand for high binding capacity and improved alkaline stability. For affinity capture of antibody Fabs, bispecific, and other variants containing a kappa light chain.
- High dynamic binding capacity protein L resin for efficient capture of antibody Fabs, bispecific, and variants containing a kappa light chain
- Stable when cleaned with 0.1 M NaOH, reducing risk for bioburden incidents
- Provides good resolution for product-related impurities in the capture of bispecific antibodies
Customers Who Bought This Also Bought