Quick-Disconnect Coupler Fittings, Hose Barbed, Straight, Dynalon®
Supplier: Dynalon
Certificates
About this item
Provides easy single-handed disconnection.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Construction from low-density polyethylene (LDPE).