Order Entry
ContactUsLinkComponent
Alto-i I Ultrapure Lab Water Polisher (18.2 MΩ-cm), Avidity Science
Alto-i I Ultrapure Lab Water Polisher (18.2 MΩ-cm), Avidity Science
Catalog #: 76509-498
Supplier:  AVIDITY SCIENCE
Alto-i I Type I water system with TOC indicator & Feed Water Conductivity
Alto-i I Ultrapure Lab Water Polisher (18.2 MΩ-cm), Avidity Science
Catalog #: 76509-498
Supplier:  AVIDITY SCIENCE

Specifications

  • Max. operating pressure:
    6 bar
  • W×D×H:
    22.6×13.2×19.4"
  • Description:
    Alto-i I TC Type I water system
  • Flow rate:
    2 L/min
  • Model:
    Alto-i I
  • Bacteria:
    <0.01 CFU/ml
  • Endotoxin:
    <0.001 EU/ml when using a TC004 POU filter
  • DNases:
    <5 pg/ml
  • Feed water:
    Reverse Osmosis (RO) or Deionized (DI)
  • RNases:
    <1 pg/ml
  • Resistivity at +25 °C:
    18.2 MΩ
  • TOC content:
    ≤5 ppb
  • Weight:
    36.4 lbs
  • Cat. no.:
    76509-498

Specifications

About this item

Provides Type 1- ultrapure water on demand for applications like cellular analysis, HPLC and mAbs.

  • Fast, volumetric and variable dispense up to 2 L/min
  • Ability to dispense Type 1 water
  • Dual wavelength ¹⁸⁵/₂₅₄ nm UV lamp
  • Data capture with printable option
  • Easy to change consumables with traceability and data tags
  • Interactive 7" touch screen
  • Continuous TOC and water quality monitoring
  • Volumetric and drop by drop dispensing
  • Color coded and adjustable dispensing stations with up to 2 remote dispensing options
  • Fed by pretreated RO or DI water

Alto-i provides 18.2 MΩ-cm water on demand. The system is fed from a pretreated supply and produces Type 1 water instantly, as required. The integrated dispenser or optional remote dispenser provides Type 1 water up to 2 liters per minute. Alto-i has quick-changing, long-lasting consumables provided in the start up package. Total Organic Carbon (TOC) monitoring is critical when organics will affect results, so a real time TOC reading provides absolute confidence in your water.

Application: Alto-i is suitable for Type 1 water usage. Type 1 water is suitable for molecular biology, electrophoresis, cell and tissue culture, sequencing, HPLC, genomics, pharmacology, monoclonal antibodies (mAbs) and any other senstive applications.

Certifications: Manufactured to ISO 9001:2015 standard. Certified to cUL, and IEC 61010-1:2010+A1.
Ordering information: Endotoxin and Nuclease-free point of use filter available (see 76337-284).
Delivery: First round of consumables are included with purchase of unit. Ships within 2 to 3 weeks and installed within 4 to 6 weeks from receiving the PO.
Customers Who Bought This Also Bought