GenScript® GenBox Mini Electrophoresis Tank, GenScript
Catalog #: 76510-506
Supplier: Genscript
Specifications
- Description:GenBox mini blot system
- Cat. no.:76510-506
Specifications
About this item
GenBox Gel electrophoresis system is designed to run 1 or 2 mini gels at the same time. GenBox mini tank is designed for ultimate convenience and consistent performance compared to traditional electrophoresis tanks. Discover the versatile and easy to use GenBox mini tank for your gel electrophoresis and blotting needs.
- Robust: Achieve reproducible protein separation with GenBox
- Convenience: Run up to 2 mini gels
- Simple: Easy to set up and operate
- Efficient: Compatible with Genbox mini blot module for traditional wet transfer
The GenBox mini electrophoresis tank runs both GenScript's 10*8 and 10*10 precast gels interchangeably. It can run one or two mini gels. The mini tank is compatible with GenBox blot modules for tank blotting. The GenBox Mini Trans-Blot module accommodates two gel holder cassettes, which is useful for blotting 2 mini PAGE gels at the same time.
Customers Who Bought This Also Bought