Specifications
- Size:100 tests
Specifications
About this item
Direct Assays: NADP+/NADPH concentrations and ratios in cell or tissue extracts.
- Sensitive and accurate: Detection limit 0.1 µM, linearity up to 10 µM NADP+/NADPH in 96-well plate assay
- Convenient: The procedure involves adding a single working reagent, and reading the optical density at time zero and 30 min at room temperature
- High-throughput: Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day
- Method: Colorimetric Determination at 565 nm
- Samples: Cells, Tissue
- Species: human, mouse, rat and more
Pyridine nucleotides play an important role in metabolism and, thus, there is continual interest in monitoring their concentration levels. Quantitative determination of NADP+/NADPH has applications in research pertaining to energy transformation and redox state of cells or tissue.
Simple, direct and automation-ready procedures for measuring NADP+/NADPH concentration are very desirable. BioAssay Systems' EnzyChromTM NADP+ /NADPH assay kit is based on a glucose dehydrogenase cycling reaction, in which the formed NADPH reduces a formazan (MTT) reagent. The intensity of the reduced product color, measured at 565 nm, is proportionate to the NADP+/NADPH concentration in the sample. This assay is highly specific for NADP+/NADPH and is not interfered by NAD+/NADH. Our assay is a convenient method to measure NADP, NADPH and their ratio.