Mini LABOPORT™ UN816KTP Diaphragm Vacuum Pumps, KNF
Supplier: KNF Neuberger Inc.
Certificates
About this item
UN816 diaphragm pumps are double-head, dry-running devices used in a wide range of laboratory applications. They transfer and pump down without contamination. The heart of these very compact pumps is a KNF structured diaphragm.
- Oil-free
- Low maintenance
- Small footprint; requires little bench space
- Available as stand alone or with vacuum gauge/regulator assembly
- Ideal for solvent filtration and degassing, and vacuum desiccation
No metal or coated-metal parts within the Ryton™ pumpheads; prevents pitting and corroding. Structured PTFE diaphragms supply consisent, reliable vacuum levels; not dependent on water pressure or temperature. Kalrez™ valves exhibit high tolerance to occasional liquid condensate, so a trap is not required except in those applications where large volumes of liquid are anticipated.
Specifications
- Catalog No:
- 82004-054
- 82004-040
- 82004-042
- 82004-044
- Model:
- N816.3KTP
- N816.3KT.45P
- N816.1.2KTP
- N816.12KT45P
- Vacuum:
- 15 torr
- 15 torr
- 120 torr
- 120 torr
- Protection class:
- IP 20 enclosure rating
- IP 20 enclosure rating
- IP 20 enclosure rating
- IP 20 enclosure rating
- W×D×H:
- 361×90×141 mm
- 361×90×141 mm
- 361×102×141 mm
- 361×102×141 mm