Mini LABOPORT™ UN811 Vacuum Diaphragm Pumps, KNF
Supplier: KNF Neuberger Inc.
Certificates
About this item
UN811 diaphragm pumps are single head, dry running devices used in a wide range of laboratory applications. They transfer, compress and pump down without contamination. The heart of these very compact pumps is a KNF structured diaphragm.
- Oil-free
- Low maintenance
- Small footprint; requires little bench space
- Available as stand alone or with vacuum gauge/regulator assembly
- Available with a dampened analog vacuum gauge and a fine regulating valve to provide simple adjustment of system vacuum levels
- For bottletop filtration, disposable filtration assemblies with collection flasks >500 ml; glass filtration assemblies with flasks <1 L; and to replace one aspirator
No metal or coated-metal parts within the Ryton™ pumpheads; prevents pitting and corroding. Structured PTFE diaphragms supply consisent, reliable vacuum levels; not dependent on water pressure or temperature. Multiport Viton™ valves exhibit high tolerance to occasional liquid condensate, so a trap is not required except in those applications where large volumes of liquid are anticipated.
Specifications
- Catalog No:
- 14224-116
- 26678-052
- Model:
- N811KVP
- N811KV.45P
- Vacuum:
- 75 torr
- 75 torr
- Protection class:
- IP 20 enclosure rating
- IP 20 enclosure rating
- W×D×H:
- 187×89.9×171.5 mm
- 187×89.9×171.5 mm